9-Anthrealdehyde 99

Supplier: ADIPOGEN CORP MS

Synonyms: 9-Anthracenecarbaldehyde, 9-Anthracenecarboxaldehyde, 9-Anthraldehyde

CDX-A0180-G005 CDX-A0180-G500
102989-230EA 19.38 USD
102989-230 102989-232
9-Anthrealdehyde 99
9-Anthrealdehyde

Fluorescent reagents (probe, stain, label) with reactive functional group for chromatography or spectroscopy. Derivatization agent. Used for membrane research, luminescent detection, molecular biology, protein modifications, protein structural analysis and proteomics. Carbohydrate labels for chromatography.

Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 369
Storage Temperature: Refrigerator
MDL Number: MFCD00001254
CAS Number: 642-31-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (HPLC)
Appearance/Color Light brown powder.
Solubility Soluble in toluene.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR