Quercetin dihydrate

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate

AG-CN2-0409-G001 AG-CN2-0409-G005
102988-164EA 34.62 USD
102988-164 102988-166
Quercetin dihydrate
Quercetin dihydrate

Multipotent flavonoid antioxidant. Potent free radical scavenger. Neuroprotective. Anticancer compound with chemosensitizing activity. Cell cycle arrest, apoptosis and autophagy inducer. Proteasome inhibitor. Pleiotropic kinase inhibitor, including tyrosine protein kinase (Trk), mitochondrial ATPase, cAMP- and cGMP-phosphodiesterases, PI3-kinase activity, phospholipase A2 and protein kinase C (PKC). DNA topoisomerases inhibitor. SIRT1 activator. Heat shock proteins inhibitor. Reversible fatty acid synthase inhibitor. Antithrombotic, antihistaminic and anti-inflammatory agent. Monoamine oxidase inhibitor. Vasodilatory compound. anti-diabetic compound.

Formula: C₁₅H₁₄O₉
MW: 338.268 g/mol
MDL Number: MFCD00149487
CAS Number: 6151-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR