n-Dodecyl-β-D-maltoside, ultrapure

Supplier: ADIPOGEN CORP MS

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside

AG-CC1-0004-G001 AG-CC1-0004-G005 AG-CC1-0004-G025
102987-822EA 166.15 USD
102987-822 102987-824 102987-826
n-Dodecyl-β-D-maltoside, ultrapure
n-Dodecyl-β-D-maltoside

Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. For the stabilization and activation of enzymes.

Formula: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR