ApoE ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT134-0001
75786-648EA 868 USD
75786-648
ApoE ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ApoE ELISA kit enabling detection of ApoE in serum, plasma, cell culture supernates, and tissue homogenates in 3.5 hours.
  • Sensitive measurement of ApoE, detecting as little as 14.063 pg/ml
  • Negligible reactivity to similar proteins
  • High throughput with results in 3.5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of ApoE in serum, plasma, cell culture supernates and tissue homogenates in human samples.
The ApoE ELISA kit is a colorimetric immunometric immunoassay kit with results in 3.5 hrs.
Ordering information: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Conjugate Buffer Substrate, and Stop Solution.