Bovine Serum Albumin (BSA)

Supplier: Equitech-Bio

BAH70-0050 BAH70-0100 BAH70-0500
103219-892EA 540.35 USD
103219-892 103219-894 103219-896
Bovine Serum Albumin (BSA)
Proteins and Peptides

With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications.


  • Reduce shear in bioreactive systems
  • Bind non-specific toxic compounds
  • Serological diluent
  • Molecular weight standard
  • Tissue culture growth nutrient


Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Our albumins are especially useful for creating defined serum-free or serum-reduced mixtures from basal media, reducing shear in bioreactor systems, and binding non-specific toxic compounds. Equitech-Bio offers albumins as a powder as well as a sterile solution. All of Equitech-Bio’s BSA is certified US origin.


Can be used as a serological diluent, as a molecular weight standard, and as a growth nutrient in tissue culture. Tested negative for Blue Tongue and Vesicular Stomatitis.


Ash content: ≤2.0%
Heavy metals: ≤20 ppm
Moisture: ≤5.0%

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR