Buffered Peptone Water
Supplier: HARDY DIAGNOSTICS
U142 U143
89407-426PK 287.09 USD
89407-426 89407-434
Buffered Peptone Water
Media for Microbiology Dehydrated Media for Microbiology
A preenrichment used for increasing the recovery of injured Salmonella spp
A preenrichment used for increasing the recovery of injured Salmonella spp. from food and other specimens prior to selective enrichment and isolation.