Bovine Blood Agar, 5%
Supplier: HARDY DIAGNOSTICS
A189 A188 A19BX
10118-938PK 46.24 USD
10118-938 10153-948 75878-358
Bovine Blood Agar, 5%
Media for Microbiology Dehydrated Media for Microbiology
For detection of organisms that may cause mastitis in dairy cattle.
(TSA with 5% Triple Washed Cow Blood) For detection of organisms that may cause mastitis in dairy cattle, such as Staphylococcus aureus and Streptococcus agalactiae.