Anaerobe Nitrate Reagent B, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z135
10015-466EA 219.31 USD
10015-466
Anaerobe Nitrate Reagent B, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Anaerobe Nitrate Reagent B, Cleve's Acid for nitrate reduction test, formulated especially for anaerobic bacteria, 15mL, ship ground only
Hardy Diagnostics Anaerobe Nitrate Reagents A, B, and C are recommended for use in determining the nitrate reduction reaction of anaerobic bacteria
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
This product is to be use with Nitrate Reagent A (Cat. no. Z134) and Nitrate Reagent C (Cat. no. Z73).