Methylene blue (Loeffler)

Supplier: HARDY DIAGNOSTICS

Synonyms: Basic Blue 9Swiss BlueMethylthioninium chlorideMethylene blue chloride3,7-Bis(dimethylamino)phenothiazin-5-ium chloride

Z88
75814-024EA 27.78 USD
75814-024
Methylene blue (Loeffler)
Methylene blue

Methylene blue, Loeffler formulation, Stain is used to examine metachromic granules in C.diphtheriae


  • Bacterial and white blood cell stain
  • Enumeration of fecal leukocytes


Used to examine metachromic granules in C. diphtheriae, screening bacteria in CSF, or for enumeration of fecal leukocytes.

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
MDL Number: MFCD00150008
CAS Number: 61-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR