Ras Activation Assays, Cell Biolabs
Supplier: Cell Biolabs
STA-400 STA-400-H STA-400-K STA-400-N
102965-086EA 855.38 USD
102965-086 102965-090 102965-094 102965-098
Ras Activation Assays, Cell Biolabs
Assays Protein Assays
Cell Biolabs' Ras Activation Assays use visible agarose beads to selectively precipitate the active form of specific Ras protein of interest
Assays are available to detect specific isoforms H-Ras, K-Ras, and N-Ras, as well as a Pan-Ras assay that detects all three isoforms. The kits use a safe, non-radioactive format.
The precipitated small GTPase is then detected by Western blot using a target-specific antibody included in the kit.