Pierce™ Maleic Anhydride Activated Plates, 96-Well, Thermo Scientific
Supplier: Invitrogen
15100 15102 15110 15112
PI15100PK 294.15 USD
PI15100 PI15102 PI15110 PI15112
Pierce™ Maleic Anhydride Activated Plates, 96-Well, Thermo Scientific
Microplates Immunology Microplates and Strips
Pierce™ Maleic activated plates allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays
  • Spontaneously react with primary amines
  • Direct, simple approach to covalently attach molecules to the well surface of an EIA plate
  • Maleic anhydride maintains its integrity and availability for coupling for months under dry, room temperature conditions
Activation level: 200 µL/well
Binding capacity: ~125 pmol biotin pentylamine (328 Da)/well
The plates are useful for immobilising peptides and other ligandsMore Product Information