Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Enzo Life Sciences
SAB-220-0500
10049-454EA 309 USD
10049-454
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Immunoglobulin G (IgG) is an immunoglobulin monomer consisting of two (gamma) heavy chains and two light chains.  Each IgG molecule contains two antigen binding domains and a single effector (Fc) domain.

Species reactivity: Rat

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR