((Ring-D5)Phe6)-Somatostatin-14, Bachem
Supplier: BACHEM AMERICAS INC
4072026.0005 4072026.0001
H-7246.0005BAEA 610.59 USD
H-7246.0005BA H-7246.0001BA
((Ring-D5)Phe6)-Somatostatin-14, Bachem
Proteins and Peptides
Stable isotope-labeled Somatostatin-14 useful for pharmacokinetic/pharmacodynamic studies in combination with Bachem product H-1490 Somatostatin-14 or the corresponding generic API (Somatostatin Ph. Eur.).