AMPIGENE® qPCR Green Mix Lo-ROX, Enzo Life Sciences
Supplier: Enzo Life Sciences
AMPIGENE®
NUC103-1000 NUC103-0200
10014-632EA 615 USD
10014-632 10014-630
AMPIGENE® qPCR Green Mix Lo-ROX, Enzo Life Sciences
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Optimized dye-based qPCR.
AMPIGENE® qPCR Green Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing dye-based real-time PCR with minimal optimization. AMPIGENE® qPCR Green Mixes use a proprietary intercalating dye that does not inhibit PCR. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Green Mixes come with either Lo-ROX or Hi-ROX concentration. For Hi-ROX, see Prod. No. ENZ-NUC104. Choose the appropriate ROX concentration for the instrument.
Ordering information: Contains 2x AMPIGENE® qPCR Green Mix Lo-ROX (includes dNTPs and polymerases).