Anti-RPS6KA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-C66 600-401-C66S
RL600401C66EA 551.64 USD
RL600401C66 RL600401C66S
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPS6KA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Horse, Bovine, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR