Anti-VIM Rat Monoclonal Antibody [clone: TM71]

Supplier: Enzo Life Sciences
KAM-CC250-E
10050-130EA 452 USD
10050-130
Anti-VIM Rat Monoclonal Antibody [clone: TM71]
Antibodies
Vimentin is one of four type II intermediate filament (IF) proteins; Desmin, Glial fibrillary acidic protein (GFAP) and Peripherin being the others. Vimentin is the most widely distributed of all IF proteins. It is typically expressed in leukocytes, blood vessel endothelial cells, and mesenchymal cells such as fibroblasts. Vimentin filaments help support cellular membranes. Vimentin networks also may help keep the nucleus and other organelles in a defined place within the cell.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: TM71
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR