Universal Methylated DNA Standard, Human, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-45005-0001
10136-020EA 371 USD
10136-020
Universal Methylated DNA Standard, Human, Enzo Life Sciences
Standards Biological Standards
Methylation standard (human) to help determine efficiency of a bisulfite DNA conversion
The Universal human methylated DNA standard with the included primers can be used in conjunction with Enzo’s Blood and tissue methylation kit or the Express methylation kit. The DNA included in this set is enzymatically methylated at all cytosine positions to help assess the efficiency of the bisulfite-mediated conversion of DNA. DNA is isolated from a male human source. The primer set is designed to amplify a fragment of the human mismatch MLH1 mismatch repair gene following bisulfite treatment. The methylated cytosines comprising CG dinucleotides remain unconverted following bisulfite treatment, whereas non-methylated cytosines are converted into uracil and detected as thymine after PCR.
Universal methylated DNA standard (human), 250ng/µl in TE buffer, pH 8.0.Control primers 20µM each primer in TE buffer, pH 8.0
Ordering information: Contains Control primers: (hMLH1 (F) and hMLH1 (R)) 20µl each and Universal methylated DNA (human) 100µg.