HighDef® Red IHC Chromogen (AP), Enzo Life Sciences
Supplier: Enzo Life Sciences
950-140-0030
10049-756EA 140 USD
10049-756
HighDef® Red IHC Chromogen (AP), Enzo Life Sciences
Stains Microbiology Stains
High definition IHC chromogen producing brilliant red color for AP-activated development.
HighDef® red IHC chromogen (AP) is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems.
Ordering information: Contains HighDef® red IHC chromogen (AP) substrate buffer and chromogen, Empty mixing bottle.