Myeloperoxidase (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-115
89141-168EA 648 USD
89141-168
Myeloperoxidase (human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of human Myeloperoxidase of culture supernatants, nasal lavage, Lithium-heparin plasma, sputum supernatant, and urine.
The Myeloperoxidase (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Tihs sensitive kit lets you quantitate as little as 19pg/ml MPO in a wide variety of sample matrices that have been validated in-house. Convenient ready-to-use liquid color-coded reagents reduce error and save time in the lab.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 13, Assay buffer 31 concentrate, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.