Complement C3a des Arg (Human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-058
89141-108EA 747 USD
89141-108
Complement C3a des Arg (Human), ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of Complement C3a des arg in human EDTA plasma.
The Complement C3a des Arg (human), EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405nm. This kit comes ready-to-use with sample preparation reagents included. The Complement C3a des Arg (human), EIA kit is specific, offering low cross-reactivity to structurally related molecules, and sensitive with quantification of as little as 120pg/ml Complement C3a des Arg.
Ordering information: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer 10 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Complement reagent A, Complement reagent B.