Bestatin, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215284401
0215284405
0215284410
0215284425
IC15284401EA
52
USD
IC15284401
IC15284405
IC15284410
IC15284425
Bestatin, MP Biomedicals
Proteins and Peptides
Soluble in methanol, acetic acid, DMSO. Stock solutions of 1 mM (2 mg/6.48 mL) in methanol are stable for approximately 1 month at -20 °C. Stock solutions can be diluted to 1:100 (10 µM solution) just before use. Working solutions should be made fresh each day.
Bestatin binds to cell surfaces and is an inhibitor for aminopeptidase B and leucine aminopeptidase.