Bovine Elastins (from Neck Ligament), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0210163601 0210163605 0210163625
IC10163601EA 65.57 USD
IC10163601 IC10163605 IC10163625
Bovine Elastins (from Neck Ligament), MP Biomedicals
Proteins and Peptides
Elastin is highly stable and hemocompatible protein now finding use in construction of biomaterials as a result of several strategies for its solubilization. It is a structural protein which has been used in studies of Chronic obstructive pulmonary disease (COPD). It may be used to investigate why there is a lack of repair for these proteins in patients with COPD.
Elastin is highly stable and hemocompatible protein now finding use in construction of biomaterials as a result of several strategies for its solubilization. It is a structural protein which has been used in studies of Chronic obstructive pulmonary disease (COPD). It may be used to investigate why there is a lack of repair for these proteins in patients with COPD.