Mouse Recombinant IL2
Supplier: IBI Scientific
RPM-140 RPM-146 RPM-145 RPM-143 RPM-142 RPM-141 RPM-144
10039-830PK 2114.25 USD
10039-830 10039-828 10039-824 10039-826 10039-832 10039-822 10039-834
Mouse Recombinant IL2
Proteins and Peptides
IBI recombinant proteins are expressed in a yeast system (Pichia pastoris) which promotes protein folding and post-translational modifications for greater bio-activity. IBI proteins are Endotoxin FREE and are purified without HIS-TAGS or an other TAG. Our yeast expression system has proven to produce more active proteins that more closely resemble their host form.
IBI proteins are lyophilized and can be shipped at room temperature. Proteins can be reconstituted in sterile PBS with at least 0.1% of carrier protein such as BSA or cell assay media.
The recombinant mouse protein IL-2 has a gene ID of 16183. The calculated molecular weight is 17.6kDa.