Anti-IgG Rabbit Polyclonal Antibody (ATTO 655)

Supplier: Rockland Immunochemical
610-457-C46 610-457-C46S
RL610-457-C46EA 414.78 USD
RL610-457-C46 RL610-457-C46S
Anti-IgG Rabbit Polyclonal Antibody (ATTO 655)
Antibodies
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: ATTO 655
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR