Anti-GDF15 Mouse Monoclonal Antibody [clone: 7C12.B3.F2]

Supplier: Rockland Immunochemical
200-301-B08 200-301-B08S
RL200-301-B08EA 565.22 USD
RL200-301-B08 RL200-301-B08S
Anti-GDF15 Mouse Monoclonal Antibody [clone: 7C12.B3.F2]
Antibodies
This Protein A purified antibody is suitable for ELISA and western blotting of human NAG-1 Protein.  This reagent is particularly useful to differentiate polymorphic forms of NAG-1 Protein present in human serum samples.

Recommended Dilutions: ELISA: 1:150000; Western Blot: 1:1000;

Type: Primary
Antigen: NAG-1 (H variant specific)
Clonality: Monoclonal
Clone: 7C12.B3.F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR