Human Adrenocorticotropic Hormone (1-39)
Supplier: Thermo Scientific Chemicals
J66508-EXD
J66508-MCR
AAJ66508-EXDEA
220
USD
AAJ66508-EXD
AAJ66508-MCR
Human Adrenocorticotropic Hormone (1-39)
Proteins and Peptides
Adrenocorticotropic Hormone (1-39), human
CAS: 12279-41-3
Form: Solid
MDL: MFCD00145721
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...