Saquayamycin B1 ≥80% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0382-C500 BVT-0382-M001
102513-814EA 131.54 USD
102513-814 102513-816
Saquayamycin B1 ≥80% (by HPLC)
Saquayamycin B1

Saquayamycin B1 is an Angucycline antibiotic.


  • Anti-bacterial
  • Farnesyl-protein transferase inhibitor
  • Anti-tumor compound


Anti-bacterial, anti-tumor compound. Active against Gram-positive bacteria and P388 leukaemia cells. Inhibitor of nitric oxide synthase (NOS) (shown for Saquayamycin A1).

MDL Number: MFCD01743886
CAS Number: 99260-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR