Echinomycin (from Streptomyces echinatus DSM 40013) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0267-M001 BVT-0267-M005
102513-696EA 152.31 USD
102513-696 102513-698
Echinomycin (from Streptomyces echinatus DSM 40013) ≥98% (by HPLC)
Echinomycin

Echinomycin is a powerful, selective inhibitor of nucleic acid synthesis in vitro.


  • Antibiotic
  • Apoptosis inducer
  • Anti-tumor compound


Potent hypoxia-inducible factor 1 (HIF-1) DNA binding activity inhibitor. Antibacterial, antifungal and antiviral.


White to off-white solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in acetone, DMSO, methanol, dichloromethane or ethyl acetate.

Formula: C₅₁H₆₄N₁₂O₁₂S₂
MW: 1101.3 g/mol
Storage Temperature: Freezer
CAS Number: 512-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR