Gilvocarcin V (from Streptomyces sp. Gö 3592) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0256-C100 BVT-0256-C250 BVT-0256-M001
102513-664EA 103.85 USD
102513-664 102513-666 102513-668
Gilvocarcin V (from Streptomyces sp. Gö 3592) ≥98% (by HPLC)
Gilvocarcin V

Gilvocarcin V is an antibiotic, anti-tumor compound.


  • Antibiotic
  • Weakly active against Gram-positive bacteria and fungi
  • Anti-tumor compound


Mediates a unique cross-linking reaction between DNA and histone H3 by light. Single strand scission and covalent binding to DNA after photoactivation.


Yellow solid


Stable for at least one year after receipt when stored at -20°C. Stock solutions are stable for at least three months when stored at -20°C.


Soluble in DMSO.

Storage Temperature: Freezer
CAS Number: 77879-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR