Concanamycin B (from Streptomyces species) ≥98% (by HPLC), high purity
Supplier: ADIPOGEN CORP MS
Concanamycin B inhibits proliferation of mouse splenic lymphocytes.
- Antifungal and larvicidal
- Antibiotic
- Suppresses bone resorption
Exhibits similar activity as concanamycin A and concanamycin C. Specific vacuolar-type H+-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Suppresses bone resorption. Inhibits proliferation of mouse splenic lymphocytes.
White to off-white solid
After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.
Soluble in DMSO, chloroform, acetonitrile or methanol; insoluble in water.
Formula:
C₄₅H₇₃NO₁₄ Storage Temperature: Freezer |
CAS Number:
81552-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...