Kirromycin (from Streptomyces collinus) ≥96% (by HPLC)
Supplier: ADIPOGEN CORP MS
Kirromycin is a member of the elfermycin group.
- Antibiotic
- Protein biosynthesis inhibitor
- Animal growth promoter
Antibiotic. Member of the elfermycin group. Antibacterial. Protein biosynthesis inhibitor. Specifically interacts with the bacterial elongation factor EF-Tu and prevents the release of EF-Tu from the bacterial ribosome after GTP hydrolysis. In the absence of ribosomes it induces EF-Tu-dependent hydrolysis of GTP. Animal growth promoter used in poultry breeding.
Yellow powder
Stable for at least one year after receipt when stored at -20°C. Store solutions at -20°C in the dark.
Soluble in methanol or DMSO.
Storage Temperature:
Freezer |
CAS Number:
50935-71-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...