Acetomycin (from Streptomyces ramulosus) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0150-M001 BVT-0150-M005
102513-560EA 131.54 USD
102513-560 102513-562
Acetomycin (from Streptomyces ramulosus) ≥98% (by HPLC)
Acetomycin

Acetomycin is a cytotoxic against several tumor cell lines in vitro.


  • Antibiotic
  • Cytotoxic
  • Anti-bacterial, anti-fungal


Antibiotic. Antibacterial, antifungal and antiprotozoal. Cytotoxic against several tumor cell lines in vitro.


White to off-white crystalline solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or methanol.

Formula: C₁₀H₁₄O₅
Storage Temperature: Freezer
CAS Number: 510-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR