Chrysomycin B (from Streptomyces species) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0100-C250 BVT-0100-M001
102513-552EA 193.85 USD
102513-552 102513-554
Chrysomycin B (from Streptomyces species) ≥98% (by HPLC)
Chrysomycin B

Chrysomycin B is a Topoisomerase II inhibitor.


  • Antibiotic
  • Less potent than chrysomycin A
  • Anti-neoplastic


Mediates a unique cross-linking reaction between DNA and histidine H3. Anti-neoplastic. Photosensitizing compound. Antitumor compound against human cell lines.


Yellow to greenish. solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or dimethylformamide; slightly soluble in 100% ethanol or methanol.

Storage Temperature: Freezer
CAS Number: 83852-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR