Lysolipin I (from Streptomyces violaceoniger) ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Lysolipin I is a glycopeptide synthesis inhibitor.
- Antibiotic
- Cytotoxic
- Antibacterial, antifungal and anticoccidial
Antibiotic. Glycopeptide synthesis inhibitor. Antibacterial, antifungal and anticoccidial. Cytotoxic.
Yellow Needles
After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.
Soluble in methylene chloride, DMSO or chloroform.
Formula:
C₂₉H₂₄ClNO₁₁ Storage Temperature: Freezer |
CAS Number:
59113-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...