Mensacarcin (from Streptomyces bottropensis) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0028-M001 BVT-0028-M005
102513-466EA 180 USD
102513-466 102513-468
Mensacarcin (from Streptomyces bottropensis) ≥98% (by HPLC)
Mensacarcin

White to off-white solid


Antibiotic. Antitumor compound. Cytotoxic.


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or methanol.

Formula: C₂₁H₂₄O₉
Storage Temperature: Freezer
CAS Number: 808750-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR