Kendomycin (from Streptomyces violaceoruber) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0001-C100 BVT-0001-C500
102513-426EA 96.92 USD
102513-426 102513-428
Kendomycin (from Streptomyces violaceoruber) ≥98% (by HPLC)
Kendomycin

Yellow powder


Potent endothelin receptor antagonist. Anti-osteoporotic. Antibacterial Cytotoxic. Mediates its cytotoxic effects, at least in part, through proteasome inhibition.


After reconstitution protect from light at -20°C. Stable for at least 1 year after receipt when stored at -20°C.

Formula: C₂₉H₄₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00076711
CAS Number: 183202-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR