Shz-1 ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-3502-M001 AG-CR1-3502-M005
102516-218EA 41.54 USD
102516-218 102516-220
Shz-1 ≥95% (by NMR)
Shz-1

Enhancer of myocardial regenerative repair by stem cells. Potent inducer of Nkx2.5 and a subset of other cardiac markers, including myocardin, and sarcomeric-tropomyosin (SalphaTM) in a variety of different stem/progenitor cells.

Formula: C₁₃H₁₁BrN₂O₃S
Storage Temperature: Freezer
MDL Number: MFCD00853265
CAS Number: 326886-05-9

Order Now

Specification Test Results

Purity 95% (NMR)
Appearance/Color White to yellow solid.
Solubility Soluble in DMSO (at least 5 mg/ml).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR