1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: ODQ

AG-CR1-3500-M010 AG-CR1-3500-M050
102516-212EA 83.08 USD
102516-212 102516-214
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥98% (by NMR)
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one

Potent and highly selective, irreversible inhibitor of soluble guanylyl cyclase (sGC). The binding is competitive with nitric oxide (NO). Tool to elucidate the nitric oxide (NO)-cGMP pathway. Apoptosis inhibitor Inhibits growth and migration of prostate cancer cells independent of its effects on GMP levels.

Formula: C₉H₅N₃O₂
MW: 187.16 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00792620
CAS Number: 41443-28-1

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to pale yellow solid.
Solubility Soluble in DMSO (at least 5 mg/ml), ethyl acetate or 100% ethanol (1.2 mg/ml); insoluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR