PD 98059 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-0118-M001 AG-CR1-0118-M005 AG-CR1-0118-M010 AG-CR1-0118-M050
102516-180EA 20.77 USD
102516-180 102516-182 102516-184 102516-186
PD 98059 ≥98% (by NMR)
PD 98059

Highly selective, reversible and cell permeable MEK (MAP kinase kinase) inhibitor. Blocks the phosphorylation and activation of the MAP kinase pathway. T cell activation inhibitor. Inhibits cell growth and cell proliferation of several cancer cells.

Formula: C₁₆H₁₃NO₃
Storage Temperature: Freezer
MDL Number: MFCD00671789
CAS Number: 167869-21-8

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Yellow solid.
Solubility Soluble in anhydrous DMSO, methanol or dichloromethane. Slightly soluble in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR