Anacardic acid ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 2-Hydroxy-6-pentadecylbenzoic acid

AG-CR1-0046-M005 AG-CR1-0046-M025
102516-060EA 62.31 USD
102516-060 102516-062
Anacardic acid ≥98%
Anacardic acid

Potent histone acetyltransferase (HAT) inhibitor. Antibacterial. SUMOylation inhibitor. Lipoxygenase inhibitor. NF-kappaB inhibitor. Anti-tumor compound. Anti-inflammatory. Apoptosis inducer. Antiproliferative.

Formula: C₂₂H₃₆O₃
MW: 348.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368254
CAS Number: 16611-84-0

Order Now

Specification Test Results

Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in 100% ethanol, methanol, DMSO, dichloromethane or ethylacetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR