MK-571 sodium salt ≥97%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Sodium (E)-3-(((3-(2-(7-chloroquinolin-2-yl)vinyl)phenyl)((3-(dimethylamino)-3-oxopropyl)thio)methyl)thio)propanoate, MK-571 sodium
AG-CR1-0021-M005
AG-CR1-0021-M025
102515-946EA
69.23
USD
102515-946
102515-948
MK-571 sodium salt ≥97%
MK-571 sodium salt
Selective leukotriene LTD4 receptor antagonist. Potent MRP1 inhibitor. Antinociceptive.
Formula:
C₂₆H₂₆ClN₂O₃S₂·Na Storage Temperature: Freezer |
MDL Number:
MFCD11114392 CAS Number: 115103-85-0 |
Specification Test Results
Purity |
|
Appearance/Color | Light yellow solid. |
Solubility | Soluble in water (20 mg/ml), 100% ethanol (10 mg/ml), DMSO or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...