FK-866 ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (E)-N-(4-(1-Benzoylpiperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide, (E)-Daporinad

AG-CR1-0011-M001 AG-CR1-0011-M005
102515-906EA 55.38 USD
102515-906 102515-908
FK-866 ≥98%
FK-866

Nampt/visfatin inhibitor. Inhibitor of NAD+ biosynthesis. Inhibits the enzyme/substrate complex and the free enzyme (Ki = 0.4 nM and Ki = 0.3 nM, respectively). Apoptosis inducer. Autophagy inducer. Causes premature senescence. Angiogenesis inhibitor.

Formula: C₂₄H₂₉N₃O₂
MW: 391.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10565943
CAS Number: 658084-64-1

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to yellow solid. (Tends to easily liquefy.)
Solubility Soluble in 100% ethanol, dimethylformamide (40 mg/ml) or DMSO (25 mg/ml); sparingly soluble in aqueous buffers.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR