D-erythro-Dihydrosphingosine 1-phosphate ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-0005-M001 AG-CR1-0005-M005
102515-880EA 124.62 USD
102515-880 102515-882
D-erythro-Dihydrosphingosine 1-phosphate ≥98% (by NMR)
D-erythro-Dihydrosphingosine-1-phosphate

Saturated analog of sphingosine 1-phosphate (1-SP1). Ligand for sphingosine 1-phosphate (S1P/EDG) receptors. Negative control for intracellular effects of sphingosine 1-phosphate. Induces chemotaxis. Antifibrotic.

Formula: C₁₈H₄₀NO₅P
Storage Temperature: Freezer
CAS Number: 19794-97-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in methanol/THF/water (60/30/10) (0.5 mg/ml). Slightly soluble in DMSO, methanol or acetic acid.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR