Mouse Recombinant IL1B (from E. coli)
Supplier: AdipoGen Life Sciences
AG-40B-0086-3010 AG-40B-0086-C010
102515-576PK 581.54 USD
102515-576 102515-574
Mouse Recombinant IL1B (from E. coli)
Proteins and Peptides
IL-1beta is produced by activated macrophages. IL-1beta stimulates thymocyte proliferation by inducing IL-2 release, B cell maturation and proliferation and fibroblast growth factor activity. IL-1beta belongs to the IL-1 family of proteins that are involved in the inflammatory response. IL-1beta in most situations requires the inflammasome complex to be cleaved and secreted.