Anti-Sirtuin Mouse Monoclonal Antibody [clone: S6R82-2]

Supplier: Adipogen
AG-20A-0091-C050 AG-20A-0091-C100
102514-246EA 138.46 USD
102514-246 102514-248
Anti-Sirtuin Mouse Monoclonal Antibody [clone: S6R82-2]
Antibodies
Sirtuin-6 is a NAD-dependent protein deacetylase with activity towards 'Lys-9' and 'Lys-56' of histone H3. Modulates acetylation of histone H3 in telomeric chromatin during the S-phase of the cell cycle. Required for genomic stability and for normal IGF1 serum levels and normal glucose homeostasis. Modulates cellular senescence and apoptosis. Regulates the production of TNF protein.

Type:
Antigen: Sirtuin
Clonality: Monoclonal
Clone: S6R82-2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR