Anti-FTO Mouse Monoclonal Antibody [clone: FT62-6]

Supplier: Adipogen
AG-20A-0083-C050 AG-20A-0083-C100
102514-214EA 283.85 USD
102514-214 102514-216
Anti-FTO Mouse Monoclonal Antibody [clone: FT62-6]
Antibodies
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse 'fused toes' mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.

Type:
Antigen: FTO
Clonality: Monoclonal
Clone: FT62-6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR