Anti-Resistin Mouse Monoclonal Antibody [clone: RRES 07]

Supplier: Adipogen
AG-20A-0015-C050 AG-20A-0015-C100
102513-964EA 249.23 USD
102513-964 102513-966
Anti-Resistin Mouse Monoclonal Antibody [clone: RRES 07]
Antibodies
The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules (RELM; RELMalpha/FIZZ 1 and RELMbeta/FIZZ 2) of FIZZ (found in inflammatory zone) are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.

Type:
Antigen: Resistin
Clonality: Monoclonal
Clone: RRES 07
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR