Anti-Macrophage Marker Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ENZ-C34935
89166-088EA 342 USD
89166-088
Anti-Macrophage Marker Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgM

  • For Immunohistochemistry

Recognizes human and monkey fixed macrophages such as "tingible macrophages", interdigitating macrophages of lymph nodes or tissue macrophages. Cross-reacts with a subpopulation of endothelial cells including those of capillaries and smaller blood vessels. Does not cross-react with B or T cells.

Type: Primary
Antigen: MACROPHAGE MARKER
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR