Anti-JNK1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA272-0020 BML-SA272-0100
89162-814EA 196.2 USD
89162-814 89162-816
Anti-JNK1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human and rat JNK 1&2 dually phosphorylated at Thr183 and Tyr185. Does not cross-react with unphosphorylated JNK1&2.

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR