Anti-PARP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA253-0025 BML-SA253-0100
89162-784EA 190.75 USD
89162-784 89162-786
Anti-PARP Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat and bovine PARP. Detects a band of ~116kDa by Western blot.

Type: Primary
Antigen: PARP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR